Assay Design



Gene Transcripts

Gene Symbol
GOLGB1
Entrez Gene ID
2804
Gene Name
golgin B1
Gene Aliases
GCP, GCP372, GOLIM1
Chromosome Location
Chr.3: 121663199 - 121749908 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256486.1 NP_001243415.1 14 - 15 8837 103
NM_001256487.1 NP_001243416.1 14 - 15 8865 103
NM_001256488.1 NP_001243417.1 13 - 14 8742 103
NM_004487.4 NP_004478.3 14 - 15 8822 103
XM_005247371.4 XP_005247428.1 14 - 15 9297 103
XM_005247372.4 XP_005247429.1 14 - 15 8873 103
XM_005247373.1 XP_005247430.1 13 - 14 8689 103
XM_006713587.1 XP_006713650.1 14 - 15 8812 103
XM_006713588.1 XP_006713651.1 14 - 15 8797 103
XM_006713589.1 XP_006713652.1 14 - 15 8794 103
XM_006713590.1 XP_006713653.1 13 - 14 8689 103
XM_006713591.1 XP_006713654.1 12 - 13 8572 103
XM_011512699.2 XP_011511001.1 14 - 15 8764 103
XM_017006189.1 XP_016861678.1 14 - 15 9282 103
XM_017006190.1 XP_016861679.1 14 - 15 9449 103
XM_017006191.1 XP_016861680.1 13 - 14 9174 103
XM_017006192.1 XP_016861681.1 14 - 15 8858 103
XM_017006193.1 XP_016861682.1 14 - 15 8855 103
XM_017006194.1 XP_016861683.1 13 - 14 8750 103
XM_017006195.1 XP_016861684.1 8 - 9 8719 103
GenBank mRNA AB371588.1 - 14 - 15 8810 103
AB593126.1 - 13 - 14 8716 103
D25542.1 - 14 - 15 8820 103
X75304.1 - 14 - 15 8810 103

View Full Product Details