Assay Design



Gene Transcripts

Gene Symbol
GOLGB1
Entrez Gene ID
2804
Gene Name
golgin B1
Gene Aliases
GCP, GCP372, GOLIM1
Chromosome Location
Chr.3: 121663199 - 121749908 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256486.1 NP_001243415.1 4 - 5 536 126
NM_001256487.1 NP_001243416.1 4 - 5 564 126
NM_001256488.1 NP_001243417.1 4 - 5 564 126
NM_004487.4 NP_004478.3 4 - 5 536 126
XM_005247371.4 XP_005247428.1 4 - 5 996 126
XM_005247372.4 XP_005247429.1 4 - 5 572 126
XM_005247373.1 XP_005247430.1 4 - 5 511 126
XM_006713587.1 XP_006713650.1 4 - 5 511 126
XM_006713588.1 XP_006713651.1 4 - 5 511 126
XM_006713589.1 XP_006713652.1 4 - 5 511 126
XM_006713590.1 XP_006713653.1 4 - 5 511 126
XM_006713591.1 XP_006713654.1 4 - 5 511 126
XM_011512699.2 XP_011511001.1 4 - 5 463 126
XM_017006189.1 XP_016861678.1 4 - 5 996 126
XM_017006190.1 XP_016861679.1 4 - 5 1166 126
XM_017006191.1 XP_016861680.1 4 - 5 996 126
XM_017006192.1 XP_016861681.1 4 - 5 572 126
XM_017006193.1 XP_016861682.1 4 - 5 572 126
XM_017006194.1 XP_016861683.1 4 - 5 572 126
GenBank mRNA AB371588.1 - 4 - 5 509 126
AB593126.1 - 4 - 5 538 126
AK296903.1 - 2 - 3 235 126
AK298986.1 - 2 - 3 210 126
AK300437.1 - 4 - 5 510 126
BC065301.1 - 4 - 5 435 126
D25542.1 - 4 - 5 519 126
X75304.1 - 4 - 5 524 126

View Full Product Details