Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016369.3 | NP_057453.1 | 1 - 2 | 279 | 83 |
GenBank mRNA | AF221069.1 | - | 1 - 2 | 226 | 83 |
AY358479.1 | - | 1 - 2 | 306 | 83 | |
BC142708.1 | - | 1 - 2 | 285 | 83 | |
BC146668.1 | - | 1 - 2 | 285 | 83 |