Assay Design



Gene Transcripts

Gene Symbol
CLDN18
Entrez Gene ID
51208
Gene Name
claudin 18
Gene Aliases
SFTA5, SFTPJ
Chromosome Location
Chr.3: 137998816 - 138033652 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002026.2 NP_001002026.1 1 - 2 272 105
GenBank mRNA AF349452.1 - 1 - 2 228 105
AK098474.1 - 1 - 2 272 105

View Full Product Details