Assay Design



Gene Transcripts

Gene Symbol
ALG1L2
Entrez Gene ID
644974
Gene Name
ALG1, chitobiosyldiphosphodolichol beta-mannosyltransferase like 2
Gene Aliases
-
Chromosome Location
Chr.3: 130081831 - 130098390 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136152.1 NP_001129624.1 1 - 2 204 57
GenBank mRNA BI833803.1 - 1 - 2 204 57

View Full Product Details