Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135095.1 | NP_001128567.1 | 5 - 6 | 607 | 87 |
NM_022763.3 | NP_073600.3 | 5 - 6 | 680 | 87 | |
XM_017007064.1 | XP_016862553.1 | 5 - 6 | 660 | 87 | |
GenBank mRNA | AB098597.1 | - | 4 - 5 | 565 | 87 |
AK127826.1 | - | 5 - 6 | 660 | 87 | |
AK223599.1 | - | 5 - 6 | 711 | 87 | |
AK314478.1 | - | 5 - 6 | 607 | 87 | |
AY358367.1 | - | 5 - 6 | 649 | 87 | |
BC039297.1 | - | 5 - 6 | 571 | 87 | |
BX648340.1 | - | 5 - 6 | 602 | 87 |