Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003292.2 | NP_003283.2 | 12 - 13 | 1686 | 95 |
GenBank mRNA | AK309473.1 | - | 13 - 14 | 1721 | 95 |
BC016514.1 | - | 12 - 13 | 1560 | 95 | |
BC025177.1 | - | 7 - 8 | 742 | 95 | |
BC094764.1 | - | 12 - 13 | 1615 | 95 | |
U69668.1 | - | 12 - 13 | 1477 | 95 | |
X63105.1 | - | 12 - 13 | 1686 | 95 | |
X66397.1 | - | 12 - 13 | 1686 | 95 |