Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003292.2 | NP_003283.2 | 9 - 10 | 1254 | 89 |
GenBank mRNA | AK309473.1 | - | 10 - 11 | 1289 | 89 |
BC016514.1 | - | 9 - 10 | 1128 | 89 | |
BC025177.1 | - | 4 - 5 | 310 | 89 | |
BC094764.1 | - | 9 - 10 | 1183 | 89 | |
U69668.1 | - | 9 - 10 | 1045 | 89 | |
X63105.1 | - | 9 - 10 | 1254 | 89 | |
X66397.1 | - | 9 - 10 | 1254 | 89 |