Assay Design



Gene Transcripts

Gene Symbol
TPR
Entrez Gene ID
7175
Gene Name
translocated promoter region, nuclear basket protein
Gene Aliases
-
Chromosome Location
Chr.1: 186311652 - 186375325 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003292.2 NP_003283.2 34 - 35 5212 79
GenBank mRNA U69668.1 - 34 - 35 5003 79
X66397.1 - 34 - 35 5212 79

View Full Product Details