Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030806.3 | NP_110433.1 | 4 - 5 | 738 | 111 |
GenBank mRNA | AF312864.1 | - | 4 - 5 | 660 | 111 |
AK056487.1 | - | 4 - 5 | 474 | 111 | |
AK312062.1 | - | 3 - 4 | 375 | 111 | |
AY258286.1 | - | 4 - 5 | 651 | 111 | |
BC028567.2 | - | 4 - 5 | 696 | 111 |