Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005076.3 | NP_005067.1 | 3 - 4 | 486 | 91 |
XM_017002198.1 | XP_016857687.1 | 2 - 3 | 227 | 91 | |
XM_017002199.1 | XP_016857688.1 | 4 - 5 | 741 | 91 | |
GenBank mRNA | AK289985.1 | - | 2 - 3 | 227 | 91 |
BC129986.1 | - | 2 - 3 | 252 | 91 | |
CR749495.1 | - | 3 - 4 | 1122 | 91 | |
X67734.1 | - | 3 - 4 | 398 | 91 | |
X68274.1 | - | 3 - 4 | 392 | 91 |