Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004947.4 | NP_004938.1 | 14 - 15 | 1275 | 126 |
XM_005264914.3 | XP_005264971.1 | 14 - 15 | 1547 | 126 | |
XM_005264915.3 | XP_005264972.1 | 14 - 15 | 1547 | 126 | |
XM_005264916.3 | XP_005264973.1 | 14 - 15 | 1325 | 126 | |
XM_005264917.3 | XP_005264974.1 | 14 - 15 | 1547 | 126 | |
XM_005264918.3 | XP_005264975.1 | 14 - 15 | 1324 | 126 | |
XM_006713008.3 | XP_006713071.1 | 14 - 15 | 1547 | 126 | |
XM_006713009.3 | XP_006713072.1 | 14 - 15 | 1547 | 126 | |
XM_006713010.3 | XP_006713073.1 | 14 - 15 | 1310 | 126 | |
XM_011533441.2 | XP_011531743.1 | 3 - 4 | 2156 | 126 | |
XM_017005825.1 | XP_016861314.1 | 14 - 15 | 1326 | 126 | |
XM_017005826.1 | XP_016861315.1 | 14 - 15 | 1316 | 126 | |
XM_017005827.1 | XP_016861316.1 | 5 - 6 | 467 | 126 | |
GenBank mRNA | AB002297.1 | - | 9 - 10 | 883 | 126 |
AY145303.1 | - | 14 - 15 | 1252 | 126 | |
AY254099.1 | - | 14 - 15 | 1275 | 126 |