Assay Design
Gene Transcripts
- Gene Symbol
- RAD51D
- Entrez Gene ID
-
5892
- Gene Name
- RAD51 paralog D
- Gene Aliases
-
BROVCA4, R51H3, RAD51L3, TRAD
- Chromosome Location
-
Chr.17: 35099792 - 35119869 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AB018362.1 |
- |
3 - 4 |
348 |
79 |
View Full Product Details