Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014460.3 | NP_055275.1 | 2 - 3 | 471 | 84 |
GenBank mRNA | AB027011.1 | - | 1 - 2 | 194 | 84 |
AK313407.1 | - | 2 - 3 | 428 | 84 | |
AL834417.1 | - | 2 - 3 | 356 | 84 | |
BC067113.1 | - | 2 - 3 | 417 | 84 | |
CR456457.1 | - | 1 - 2 | 206 | 84 |