Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007208.3 | NP_009139.1 | 5 - 6 | 767 | 89 |
GenBank mRNA | AK296626.1 | - | 6 - 7 | 817 | 89 |
AK301377.1 | - | 5 - 6 | 601 | 89 | |
BC003375.2 | - | 5 - 6 | 666 | 89 | |
BI490082.1 | - | 5 - 6 | 736 | 89 | |
CR456720.1 | - | 5 - 6 | 572 | 89 | |
X06323.1 | - | 5 - 6 | 648 | 89 |