Assay Design
Gene Transcripts
- Gene Symbol
- CCDC39
- Entrez Gene ID
-
339829
- Gene Name
- coiled-coil domain containing 39
- Gene Aliases
-
CILD14, FAP59
- Chromosome Location
-
Chr.3: 180614008 - 180679495 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AL122120.1 |
- |
4 - 5 |
533 |
107 |
- Gene Symbol
- LOC101928882
- Entrez Gene ID
-
101928882
- Gene Name
- uncharacterized LOC101928882
- Gene Aliases
-
-
- Chromosome Location
-
Chr.3: 180707589 - 180870178 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_109986.1 |
|
13 - 14 |
1550 |
107 |
GenBank mRNA |
BC047103.1 |
- |
13 - 14 |
1550 |
107 |
BC094711.1 |
- |
3 - 4 |
114 |
107 |
View Full Product Details