Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135752.1 | NP_001129224.1 | 4 - 5 | 651 | 108 |
NM_001135753.1 | NP_001129225.1 | 4 - 5 | 651 | 108 | |
NM_007265.2 | NP_009196.1 | 4 - 5 | 651 | 108 | |
NR_024203.1 | 3 - 4 | 433 | 108 | ||
GenBank mRNA | AK022616.1 | - | 4 - 5 | 599 | 108 |
AK225519.1 | - | 4 - 5 | 627 | 108 | |
AK315711.1 | - | 4 - 5 | 624 | 108 | |
BC000721.2 | - | 4 - 5 | 615 | 108 | |
CR457407.1 | - | 3 - 4 | 408 | 108 | |
D88208.1 | - | 4 - 5 | 615 | 108 |