Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152635.2 | NP_689848.1 | 6 - 7 | 1192 | 76 |
GenBank mRNA | AK096435.1 | - | 6 - 7 | 1167 | 76 |
AY013707.2 | - | 6 - 7 | 1023 | 76 | |
AY358339.1 | - | 6 - 7 | 1145 | 76 |