Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001276373.1 | NP_001263302.1 | 9 - 10 | 823 | 99 |
NM_007122.4 | NP_009053.1 | 9 - 10 | 919 | 99 | |
NM_207005.2 | NP_996888.1 | 9 - 10 | 888 | 99 | |
GenBank mRNA | AB098540.1 | - | 7 - 8 | 681 | 99 |
AK314876.1 | - | 9 - 10 | 839 | 99 | |
AL832119.1 | - | 9 - 10 | 956 | 99 | |
BC035505.2 | - | 9 - 10 | 867 | 99 | |
KU178379.1 | - | 7 - 8 | 712 | 99 | |
KU178380.1 | - | 7 - 8 | 681 | 99 | |
X55666.1 | - | 9 - 10 | 832 | 99 |