Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015054.1 | NP_055869.1 | 13 - 14 | 1979 | 80 |
XM_005268738.2 | XP_005268795.1 | 14 - 15 | 2085 | 80 | |
XM_005268739.3 | XP_005268796.1 | 12 - 13 | 1756 | 80 | |
XM_011538054.2 | XP_011536356.1 | 3 - 4 | 454 | 80 | |
XM_017019047.1 | XP_016874536.1 | 13 - 14 | 1939 | 80 | |
XM_017019048.1 | XP_016874537.1 | 13 - 14 | 1902 | 80 | |
XM_017019049.1 | XP_016874538.1 | 13 - 14 | 1939 | 80 | |
XM_017019050.1 | XP_016874539.1 | 13 - 14 | 1884 | 80 | |
XM_017019051.1 | XP_016874540.1 | 14 - 15 | 2030 | 80 | |
GenBank mRNA | AB014601.2 | - | 9 - 10 | 1420 | 80 |
AK026472.1 | - | 4 - 5 | 576 | 80 | |
AK125813.1 | - | 13 - 14 | 1939 | 80 | |
AL136821.1 | - | 6 - 7 | 784 | 80 | |
AL833850.1 | - | 3 - 4 | 433 | 80 | |
BC127017.1 | - | 13 - 14 | 1797 | 80 | |
BX647380.1 | - | 7 - 8 | 950 | 80 |