Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270520.1 | NP_001257449.1 | 13 - 14 | 1709 | 60 |
NM_014992.2 | NP_055807.1 | 12 - 13 | 1603 | 60 | |
XM_005267430.1 | XP_005267487.1 | 13 - 14 | 1761 | 60 | |
XM_005267431.1 | XP_005267488.1 | 13 - 14 | 1861 | 60 | |
GenBank mRNA | AB014566.1 | - | 12 - 13 | 1589 | 60 |
AK291790.1 | - | 13 - 14 | 1678 | 60 | |
BC024781.1 | - | 3 - 4 | 350 | 60 | |
BC038428.1 | - | 13 - 14 | 1701 | 60 | |
BC064999.1 | - | 13 - 14 | 1692 | 60 |