Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270520.1 | NP_001257449.1 | 19 - 20 | 2500 | 69 |
NM_014992.2 | NP_055807.1 | 19 - 20 | 2424 | 69 | |
XM_005267430.1 | XP_005267487.1 | 20 - 21 | 2582 | 69 | |
XM_005267431.1 | XP_005267488.1 | 20 - 21 | 2682 | 69 | |
GenBank mRNA | AB014566.1 | - | 19 - 20 | 2410 | 69 |
AK021890.1 | - | 9 - 10 | 848 | 69 | |
AK291790.1 | - | 19 - 20 | 2469 | 69 | |
BC024781.1 | - | 9 - 10 | 1141 | 69 | |
BC038428.1 | - | 19 - 20 | 2492 | 69 | |
BC064999.1 | - | 19 - 20 | 2483 | 69 |