Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193493.1 | NP_001180422.1 | 2 - 3 | 215 | 74 |
NM_003038.4 | NP_003029.2 | 2 - 3 | 849 | 74 | |
XM_006712079.2 | XP_006712142.1 | 2 - 3 | 215 | 74 | |
GenBank mRNA | AK126636.1 | - | 1 - 2 | 2039 | 74 |
AK223486.1 | - | 2 - 3 | 811 | 74 | |
AK295687.1 | - | 2 - 3 | 170 | 74 | |
AK298472.1 | - | 3 - 4 | 760 | 74 | |
AK313052.1 | - | 2 - 3 | 813 | 74 | |
AK315986.1 | - | 3 - 4 | 771 | 74 | |
BC026216.1 | - | 2 - 3 | 750 | 74 | |
BC072423.1 | - | 2 - 3 | 809 | 74 | |
DA218921.1 | - | 2 - 3 | 217 | 74 | |
L14595.1 | - | 2 - 3 | 749 | 74 | |
L19444.1 | - | 2 - 3 | 728 | 74 |