Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001306079.1 | NP_001293008.1 | 31 - 32 | 5223 | 80 |
NM_004341.4 | NP_004332.2 | 32 - 33 | 5412 | 80 | |
XM_005264555.3 | XP_005264612.1 | 32 - 33 | 5545 | 80 | |
XM_006712101.2 | XP_006712164.1 | 31 - 32 | 5356 | 80 | |
GenBank mRNA | AK226105.1 | - | 31 - 32 | 5205 | 80 |
BC014178.2 | - | 7 - 8 | 1129 | 80 | |
BC065510.1 | - | 32 - 33 | 5412 | 80 | |
BG766243.1 | - | 4 - 5 | 588 | 80 | |
BU558264.1 | - | 3 - 4 | 414 | 80 | |
D78586.1 | - | 32 - 33 | 5276 | 80 |