Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013943.2 | NP_039234.1 | 5 - 6 | 882 | 83 |
GenBank mRNA | AF097330.1 | - | 5 - 6 | 737 | 83 |
AF109196.1 | - | 5 - 6 | 675 | 83 | |
AK001624.1 | - | 5 - 6 | 769 | 84 | |
AK095959.1 | - | 5 - 6 | 703 | 83 | |
AK301466.1 | - | 5 - 6 | 680 | 83 | |
AK307142.1 | - | 3 - 4 | 447 | 83 | |
AK315051.1 | - | 5 - 6 | 719 | 83 | |
AL117424.1 | - | 5 - 6 | 794 | 83 | |
BC012444.1 | - | 5 - 6 | 747 | 83 | |
CR533501.1 | - | 5 - 6 | 597 | 83 |