Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016618.2 | NP_057702.1 | 2 - 3 | 286 | 136 |
XM_017004292.1 | XP_016859781.1 | 2 - 3 | 587 | 136 | |
GenBank mRNA | AF208845.1 | - | 2 - 3 | 214 | 136 |
AF217520.1 | - | 2 - 3 | 225 | 136 | |
BC107580.1 | - | 2 - 3 | 252 | 136 |