Assay Design



Gene Transcripts

Gene Symbol
FBP1
Entrez Gene ID
2203
Gene Name
fructose-bisphosphatase 1
Gene Aliases
FBP
Chromosome Location
Chr.9: 94603133 - 94640257 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000507.3 NP_000498.2 2 - 3 655 103
NM_001127628.1 NP_001121100.1 3 - 4 554 103
XM_006717005.3 XP_006717068.1 2 - 3 368 103
GenBank mRNA AF073475.1 - 2 - 3 321 103
AK223395.1 - 2 - 3 599 103
AK292199.1 - 3 - 4 593 103
AK316583.1 - 2 - 3 656 103
AY544122.1 - 2 - 3 411 103
BC012927.1 - 2 - 3 521 103
D26054.1 - 2 - 3 330 103
D26055.1 - 2 - 3 330 103
D26056.1 - 2 - 3 330 103
L10320.1 - 2 - 3 466 103
M19922.1 - 2 - 3 522 103

View Full Product Details