Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005233.5 | NP_005224.2 | 4 - 5 | 1199 | 94 |
NM_182644.2 | NP_872585.1 | 4 - 5 | 1199 | 94 | |
XM_005264715.2 | XP_005264772.1 | 4 - 5 | 1124 | 94 | |
XM_005264716.2 | XP_005264773.1 | 4 - 5 | 1194 | 94 | |
GenBank mRNA | AF213459.1 | - | 4 - 5 | 1199 | 94 |
AF213460.1 | - | 4 - 5 | 1061 | 94 | |
AK291411.1 | - | 4 - 5 | 1032 | 94 | |
BC063282.1 | - | 4 - 5 | 1103 | 94 | |
M83941.1 | - | 4 - 5 | 1074 | 94 |