Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278511.1 | NP_001265440.1 | 15 - 16 | 2074 | 91 |
NM_004644.4 | NP_004635.2 | 16 - 17 | 2170 | 91 | |
XM_017022640.1 | XP_016878129.1 | 15 - 16 | 2302 | 91 | |
GenBank mRNA | AF022152.1 | - | 16 - 17 | 1999 | 91 |
AK097827.1 | - | 8 - 9 | 1067 | 91 | |
BC093739.1 | - | 16 - 17 | 2041 | 91 | |
BC143342.1 | - | 15 - 16 | 1945 | 91 | |
CR749860.1 | - | 15 - 16 | 2169 | 91 | |
U37673.1 | - | 16 - 17 | 2026 | 91 |