Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145345.2 | NP_663320.2 | 5 - 6 | 574 | 114 |
NM_183008.2 | NP_892120.2 | 6 - 7 | 579 | 114 | |
GenBank mRNA | AF520989.1 | - | 4 - 5 | 283 | 114 |
AF521017.1 | - | 4 - 5 | 284 | 114 | |
AF521018.1 | - | 4 - 5 | 272 | 114 | |
AK090792.1 | - | 2 - 3 | 149 | 114 | |
AK093258.1 | - | 6 - 7 | 523 | 114 | |
AK291503.1 | - | 4 - 5 | 279 | 114 | |
AK296179.1 | - | 3 - 4 | 181 | 114 | |
AY007163.1 | - | 3 - 4 | 165 | 114 | |
BC038106.1 | - | 5 - 6 | 391 | 114 | |
BX647774.1 | - | 5 - 6 | 387 | 114 | |
BX648412.1 | - | 4 - 5 | 288 | 114 | |
CR749413.1 | - | 5 - 6 | 385 | 114 | |
DA421971.1 | - | 5 - 6 | 386 | 114 | |
DA989799.1 | - | - | 490 | 88 |