Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000443.3 | NP_000434.1 | 24 - 25 | 3156 | 92 |
NM_018849.2 | NP_061337.1 | 24 - 25 | 3156 | 92 | |
NM_018850.2 | NP_061338.1 | 23 - 24 | 3015 | 92 | |
XM_011516308.2 | XP_011514610.2 | 24 - 25 | 3180 | 92 | |
XM_011516309.2 | XP_011514611.2 | 24 - 25 | 3180 | 92 | |
XM_011516310.2 | XP_011514612.2 | 23 - 24 | 3075 | 92 | |
XM_011516311.2 | XP_011514613.2 | 24 - 25 | 3051 | 92 | |
XM_011516312.2 | XP_011514614.2 | 23 - 24 | 3039 | 92 | |
XM_011516313.2 | XP_011514615.1 | 23 - 24 | 3038 | 92 | |
XM_011516315.2 | XP_011514617.2 | 19 - 20 | 2556 | 92 | |
XM_017012323.1 | XP_016867812.1 | 24 - 25 | 3167 | 92 | |
GenBank mRNA | BC042531.1 | - | 24 - 25 | 3192 | 92 |
M23234.1 | - | 24 - 25 | 3112 | 92 | |
X06181.1 | - | 4 - 5 | 515 | 92 |