Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024764.3 | NP_079040.2 | 24 - 25 | 3112 | 108 |
GenBank mRNA | AK024360.1 | - | 10 - 11 | 1640 | 108 |
AK126034.1 | - | 24 - 25 | 3096 | 108 | |
BC126214.1 | - | 24 - 25 | 3066 | 108 | |
BC126216.1 | - | 24 - 25 | 3066 | 108 |