Assay Design



Gene Transcripts

Gene Symbol
DDHD2
Entrez Gene ID
23259
Gene Name
DDHD domain containing 2
Gene Aliases
SAMWD1, SPG54, iPLA(1)gamma
Chromosome Location
Chr.8: 38231343 - 38273442 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164232.1 NP_001157704.1 14 - 15 1919 74
NM_015214.2 NP_056029.2 14 - 15 2096 74
XM_005273454.2 XP_005273511.1 14 - 15 2244 74
XM_005273455.3 XP_005273512.1 14 - 15 2244 74
XM_005273456.3 XP_005273513.1 13 - 14 2154 74
XM_011544456.1 XP_011542758.1 14 - 15 2244 74
XM_017013252.1 XP_016868741.1 14 - 15 2053 74
XM_017013253.1 XP_016868742.1 14 - 15 2244 74
XM_017013254.1 XP_016868743.1 13 - 14 1833 74
XM_017013255.1 XP_016868744.1 12 - 13 1842 74
XM_017013256.1 XP_016868745.1 13 - 14 1932 74
GenBank mRNA AB018268.1 - 11 - 12 1306 74
AK023218.1 - 10 - 11 1125 74
AK056525.1 - 14 - 15 1998 74
AK094951.1 - 3 - 4 2509 74
AK125904.1 - 14 - 15 1919 74
AK127040.1 - 5 - 6 1447 74
AK299875.1 - 12 - 13 1503 74
BC010504.1 - 9 - 10 1067 74

View Full Product Details