Assay Design



Gene Transcripts

Gene Symbol
DDHD2
Entrez Gene ID
23259
Gene Name
DDHD domain containing 2
Gene Aliases
SAMWD1, SPG54, iPLA(1)gamma
Chromosome Location
Chr.8: 38231343 - 38273442 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164232.1 NP_001157704.1 4 - 5 702 120
NM_001164234.1 NP_001157706.1 4 - 5 879 120
NM_015214.2 NP_056029.2 4 - 5 879 120
XM_005273454.2 XP_005273511.1 4 - 5 1027 120
XM_005273455.3 XP_005273512.1 4 - 5 1027 120
XM_005273456.3 XP_005273513.1 4 - 5 1027 120
XM_011544456.1 XP_011542758.1 4 - 5 1027 120
XM_017013252.1 XP_016868741.1 4 - 5 836 120
XM_017013253.1 XP_016868742.1 4 - 5 1027 120
XM_017013254.1 XP_016868743.1 4 - 5 684 120
XM_017013255.1 XP_016868744.1 4 - 5 783 120
XM_017013256.1 XP_016868745.1 4 - 5 783 120
GenBank mRNA AB018268.1 - 1 - 2 89 120
AK125904.1 - 4 - 5 702 120

View Full Product Details