Assay Design



Gene Transcripts

Gene Symbol
DDHD2
Entrez Gene ID
23259
Gene Name
DDHD domain containing 2
Gene Aliases
SAMWD1, SPG54, iPLA(1)gamma
Chromosome Location
Chr.8: 38231343 - 38273442 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164232.1 NP_001157704.1 8 - 9 1256 128
NM_015214.2 NP_056029.2 8 - 9 1433 128
XM_005273454.2 XP_005273511.1 8 - 9 1581 128
XM_005273455.3 XP_005273512.1 8 - 9 1581 128
XM_005273456.3 XP_005273513.1 7 - 8 1491 128
XM_011544456.1 XP_011542758.1 8 - 9 1581 128
XM_017013252.1 XP_016868741.1 8 - 9 1390 128
XM_017013253.1 XP_016868742.1 8 - 9 1581 128
GenBank mRNA AK023218.1 - 4 - 5 462 128
AK056525.1 - 8 - 9 1335 128
AK125904.1 - 8 - 9 1256 128
AK299875.1 - 6 - 7 931 128
BC010504.1 - 3 - 4 404 128

View Full Product Details