Assay Design



Gene Transcripts

Gene Symbol
DDHD2
Entrez Gene ID
23259
Gene Name
DDHD domain containing 2
Gene Aliases
SAMWD1, SPG54, iPLA(1)gamma
Chromosome Location
Chr.8: 38231343 - 38273442 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164232.1 NP_001157704.1 10 - 11 1447 140
NM_015214.2 NP_056029.2 10 - 11 1624 140
XM_005273454.2 XP_005273511.1 10 - 11 1772 140
XM_005273455.3 XP_005273512.1 10 - 11 1772 140
XM_005273456.3 XP_005273513.1 9 - 10 1682 140
XM_011544456.1 XP_011542758.1 10 - 11 1772 140
XM_017013252.1 XP_016868741.1 10 - 11 1581 140
XM_017013253.1 XP_016868742.1 10 - 11 1772 140
XM_017013254.1 XP_016868743.1 9 - 10 1361 140
XM_017013255.1 XP_016868744.1 8 - 9 1370 140
XM_017013256.1 XP_016868745.1 9 - 10 1460 140
GenBank mRNA AB018268.1 - 7 - 8 834 140
AK023218.1 - 6 - 7 653 140
AK125904.1 - 10 - 11 1447 140
AK127040.1 - 1 - 2 975 140
AK299875.1 - 8 - 9 1122 140
BC010504.1 - 5 - 6 595 140

View Full Product Details