Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015594.2 | NP_056409.1 | 4 - 5 | 382 | 136 |
XM_011524596.1 | XP_011522898.1 | 4 - 5 | 308 | 136 | |
GenBank mRNA | AL117394.1 | - | 5 - 6 | 2731 | 136 |
BC096718.1 | - | 4 - 5 | 382 | 136 |