Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001303451.1 | NP_001290380.1 | 1 - 2 | 138 | 62 |
NM_024731.3 | NP_079007.2 | 1 - 2 | 138 | 62 | |
GenBank mRNA | BC032400.1 | - | 1 - 2 | 114 | 62 |
BC033821.1 | - | 1 - 2 | 124 | 62 | |
DA049618.1 | - | 1 - 2 | 140 | 62 |