Assay Design



Gene Transcripts

Gene Symbol
PHLDB1
Entrez Gene ID
23187
Gene Name
pleckstrin homology like domain family B member 1
Gene Aliases
LL5A
Chromosome Location
Chr.11: 118606296 - 118658038 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144758.2 NP_001138230.1 17 - 18 3635 53
NM_001144759.2 NP_001138231.1 16 - 17 3494 53
NM_015157.3 NP_055972.1 18 - 19 3858 53
XM_005271468.3 XP_005271525.1 18 - 19 4449 53
XM_005271469.3 XP_005271526.1 18 - 19 4449 53
XM_005271470.3 XP_005271527.1 17 - 18 4404 53
XM_005271471.3 XP_005271528.1 17 - 18 4404 53
XM_005271472.3 XP_005271529.1 17 - 18 4308 53
XM_005271473.3 XP_005271530.1 16 - 17 4263 53
XM_005271474.4 XP_005271531.1 16 - 17 4263 53
XM_005277680.3 XP_005277737.1 21 - 22 4890 53
XM_006718796.3 XP_006718859.1 21 - 22 4079 53
XM_006718797.3 XP_006718860.1 20 - 21 4749 53
XM_006718798.3 XP_006718861.1 19 - 20 4578 53
XM_006718799.3 XP_006718862.1 18 - 19 4533 53
XM_006718800.3 XP_006718863.1 18 - 19 4533 53
XM_006718801.3 XP_006718864.1 18 - 19 4437 53
XM_006718802.3 XP_006718865.1 17 - 18 4392 53
XM_006718803.3 XP_006718866.1 17 - 18 4392 53
XM_011542703.2 XP_011541005.1 21 - 22 4893 53
XM_011542704.2 XP_011541006.1 21 - 22 4893 53
XM_011542705.2 XP_011541007.1 20 - 21 4848 53
XM_011542706.2 XP_011541008.1 20 - 21 4848 53
XM_011542707.2 XP_011541009.1 20 - 21 4764 53
XM_011542708.2 XP_011541010.1 21 - 22 4082 53
XM_011542709.2 XP_011541011.1 21 - 22 4392 53
XM_011542710.2 XP_011541012.1 20 - 21 4722 53
XM_011542711.2 XP_011541013.1 18 - 19 3799 53
XM_011542713.2 XP_011541015.1 18 - 19 3793 53
XM_011542715.2 XP_011541017.1 11 - 12 2770 53
XM_017017407.1 XP_016872896.1 20 - 21 4761 53
XM_017017408.1 XP_016872897.1 19 - 20 4704 53
XM_017017409.1 XP_016872898.1 19 - 20 4677 53
XM_017017410.1 XP_016872899.1 19 - 20 4620 53
GenBank mRNA AB014538.2 - 17 - 18 3612 53
AB094090.1 - 18 - 19 3858 53
AB177859.1 - 13 - 14 2784 53
AK027510.1 - 8 - 9 1123 53
AK074070.2 - 16 - 17 3443 53
AK074747.1 - 2 - 3 378 53
AK074918.1 - 10 - 11 1330 53
AK125783.1 - 19 - 20 2862 53
BC013031.2 - 10 - 11 1480 53
BC098586.1 - 16 - 17 3988 53
BX648176.1 - 11 - 12 1646 53
CR749783.1 - 17 - 18 4151 53

View Full Product Details