Assay Design
Gene Transcripts
- Gene Symbol
- PHLDB1
- Entrez Gene ID
-
23187
- Gene Name
- pleckstrin homology like domain family B member 1
- Gene Aliases
-
LL5A
- Chromosome Location
-
Chr.11: 118606296 - 118658038 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_015157.3 |
NP_055972.1 |
1 - 2 |
172 |
107 |
GenBank mRNA |
AB094090.1 |
- |
1 - 2 |
172 |
107 |
View Full Product Details