Assay Design



Gene Transcripts

Gene Symbol
PHLDB1
Entrez Gene ID
23187
Gene Name
pleckstrin homology like domain family B member 1
Gene Aliases
LL5A
Chromosome Location
Chr.11: 118606296 - 118658038 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144758.2 NP_001138230.1 22 - 23 4126 85
NM_015157.3 NP_055972.1 23 - 24 4349 85
XM_005271468.3 XP_005271525.1 23 - 24 4940 85
XM_005271470.3 XP_005271527.1 22 - 23 4895 85
XM_005271472.3 XP_005271529.1 22 - 23 4799 85
XM_005271473.3 XP_005271530.1 21 - 22 4754 85
XM_005277680.3 XP_005277737.1 26 - 27 5381 85
XM_006718796.3 XP_006718859.1 26 - 27 4570 85
XM_006718797.3 XP_006718860.1 25 - 26 5240 85
XM_006718798.3 XP_006718861.1 24 - 25 5069 85
XM_006718799.3 XP_006718862.1 23 - 24 5024 85
XM_006718801.3 XP_006718864.1 23 - 24 4928 85
XM_006718802.3 XP_006718865.1 22 - 23 4883 85
XM_011542703.2 XP_011541005.1 26 - 27 5384 85
XM_011542705.2 XP_011541007.1 25 - 26 5339 85
XM_011542707.2 XP_011541009.1 25 - 26 5255 85
XM_011542708.2 XP_011541010.1 26 - 27 4573 85
XM_011542709.2 XP_011541011.1 26 - 27 4883 85
XM_011542710.2 XP_011541012.1 25 - 26 5213 85
XM_011542711.2 XP_011541013.1 23 - 24 4290 85
XM_011542713.2 XP_011541015.1 23 - 24 4284 85
XM_017017407.1 XP_016872896.1 25 - 26 5252 85
XM_017017410.1 XP_016872899.1 24 - 25 5111 85
GenBank mRNA AB014538.2 - 22 - 23 4103 85
AB094090.1 - 23 - 24 4349 85

View Full Product Details