Assay Design



Gene Transcripts

Gene Symbol
ENAH
Entrez Gene ID
55740
Gene Name
enabled homolog (Drosophila)
Gene Aliases
ENA, MENA, NDPP1
Chromosome Location
Chr.1: 225486832 - 225660045 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017001757.1 XP_016857246.1 1 - 2 55 85

View Full Product Details