Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001195302.1 | NP_001182231.1 | 1 - 2 | 212 | 136 |
NM_001204911.1 | NP_001191840.1 | 1 - 2 | 212 | 136 | |
NM_024598.3 | NP_078874.2 | 1 - 2 | 212 | 136 | |
XM_011523330.1 | XP_011521632.1 | 1 - 2 | 178 | 136 | |
GenBank mRNA | AK023216.1 | - | 1 - 2 | 184 | 136 |
AK301494.1 | - | 1 - 2 | 162 | 136 | |
AK303121.1 | - | 1 - 2 | 167 | 136 | |
BC004415.1 | - | 1 - 2 | 148 | 136 | |
BC006291.2 | - | 1 - 2 | 138 | 136 | |
BC007774.2 | - | 1 - 2 | 142 | 136 | |
BC010099.2 | - | 1 - 2 | 165 | 136 | |
BC021554.1 | - | 1 - 2 | 132 | 136 | |
BM193105.1 | - | 1 - 2 | 185 | 136 | |
DC363309.1 | - | 1 - 2 | 213 | 136 | |
DC380401.1 | - | 1 - 2 | 212 | 136 |