Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004181.4 | NP_004172.2 | 6 - 7 | 549 | 80 |
GenBank mRNA | AK054579.1 | - | 5 - 6 | 984 | 80 |
AK315368.1 | - | 6 - 7 | 498 | 80 | |
BC000332.2 | - | 6 - 7 | 518 | 80 | |
BC005117.1 | - | 6 - 7 | 477 | 80 | |
BC006305.2 | - | 6 - 7 | 489 | 80 | |
BC018838.2 | - | 6 - 7 | 518 | 80 | |
FJ224355.1 | - | 6 - 7 | 518 | 80 | |
GQ472201.1 | - | 6 - 7 | 489 | 80 | |
X04741.1 | - | 6 - 7 | 451 | 80 |