Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012514.2 | NP_001012532.1 | 2 - 3 | 436 | 70 |
NM_001287240.1 | NP_001274169.1 | 3 - 4 | 513 | 70 | |
NM_001287241.1 | NP_001274170.1 | 4 - 5 | 709 | 70 | |
NM_030926.5 | NP_112188.1 | 3 - 4 | 577 | 70 | |
GenBank mRNA | AB003629.1 | - | 3 - 4 | 446 | 70 |
AF271781.1 | - | 3 - 4 | 562 | 70 | |
AF272043.1 | - | 3 - 4 | 559 | 70 | |
AK056321.1 | - | 2 - 3 | 383 | 70 | |
AK075361.1 | - | 3 - 4 | 542 | 70 | |
AK090975.1 | - | 3 - 4 | 488 | 70 | |
AK222697.1 | - | 3 - 4 | 524 | 70 | |
AK303199.1 | - | 3 - 4 | 531 | 70 | |
AK307702.1 | - | 2 - 3 | 345 | 70 | |
AL136603.1 | - | 3 - 4 | 518 | 70 | |
AL713651.1 | - | 3 - 4 | 501 | 70 | |
AY049777.1 | - | 3 - 4 | 463 | 70 | |
BC002424.1 | - | 3 - 4 | 504 | 70 | |
BC050668.1 | - | 2 - 3 | 371 | 70 | |
BC098563.1 | - | 3 - 4 | 490 | 70 | |
BM842614.1 | - | 3 - 4 | 566 | 70 | |
BX444331.2 | - | 3 - 4 | 512 | 70 | |
CR457208.1 | - | 3 - 4 | 446 | 70 |