Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282541.1 | NP_001269470.1 | 21 - 22 | 2046 | 137 |
NM_001282542.1 | NP_001269471.1 | 21 - 22 | 2046 | 137 | |
NM_003176.3 | NP_003167.2 | 21 - 22 | 2043 | 137 | |
NR_104211.1 | 21 - 22 | 2043 | 137 | ||
XM_005271155.3 | XP_005271212.1 | 20 - 21 | 2367 | 137 | |
XM_005271156.1 | XP_005271213.1 | 20 - 21 | 1841 | 137 | |
XM_006710859.1 | XP_006710922.1 | 21 - 22 | 1954 | 137 | |
XM_011542037.2 | XP_011540339.1 | 21 - 22 | 2010 | 137 | |
XM_011542038.2 | XP_011540340.1 | 14 - 15 | 1237 | 137 | |
XM_017002184.1 | XP_016857673.1 | 21 - 22 | 1846 | 137 | |
XM_017002185.1 | XP_016857674.1 | 15 - 16 | 1272 | 137 | |
GenBank mRNA | AF273043.1 | - | 21 - 22 | 1981 | 137 |
AK315268.1 | - | 21 - 22 | 2005 | 137 | |
BC126266.1 | - | 21 - 22 | 1920 | 137 | |
BC144019.1 | - | 21 - 22 | 1923 | 137 | |
D67035.1 | - | 21 - 22 | 1970 | 137 | |
X95654.1 | - | 21 - 22 | 1897 | 137 |