Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145450.1 | NP_001138922.1 | 3 - 4 | 309 | 122 |
GenBank mRNA | AY367766.1 | - | 3 - 4 | 309 | 122 |
BC095472.1 | - | 1 - 2 | 119 | 122 | |
BC102032.1 | - | 3 - 4 | 122 | ||
BC102034.1 | - | 3 - 4 | 206 | 122 | |
BC102035.1 | - | 3 - 4 | 206 | 122 | |
BE466470.1 | - | 2 - 3 | 122 |