Assay Design



Gene Transcripts

Gene Symbol
MGME1
Entrez Gene ID
92667
Gene Name
mitochondrial genome maintenance exonuclease 1
Gene Aliases
C20orf72, DDK1, MTDPS11, bA504H3.4
Chromosome Location
Chr.20: 17968594 - 17991122 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011529394.1 XP_011527696.1 1 - 2 92 153
XM_017028128.1 XP_016883617.1 1 - 2 351 153

View Full Product Details