Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199.3 | NP_001190.1 | 8 - 9 | 1344 | 73 |
NM_006129.4 | NP_006120.1 | 8 - 9 | 1344 | 73 | |
NR_033403.1 | 8 - 9 | 1381 | 73 | ||
NR_033404.1 | 8 - 9 | 1381 | 73 | ||
XM_006716386.2 | XP_006716449.2 | 8 - 9 | 1344 | 73 | |
XM_011544617.1 | XP_011542919.1 | 8 - 9 | 1344 | 73 | |
XM_017013738.1 | XP_016869227.1 | 8 - 9 | 1344 | 73 | |
GenBank mRNA | AB209590.1 | - | 8 - 9 | 1297 | 73 |
AK291620.1 | - | 8 - 9 | 1197 | 73 | |
BC136679.1 | - | 8 - 9 | 1160 | 73 | |
BC142953.1 | - | 8 - 9 | 1208 | 73 | |
BC144365.1 | - | 8 - 9 | 1193 | 73 | |
BC144366.1 | - | 8 - 9 | 1197 | 73 | |
M22488.1 | - | 8 - 9 | 1107 | 73 | |
U50330.1 | - | 8 - 9 | 1078 | 73 | |
Y08723.1 | - | 9 - 10 | 1250 | 73 | |
Y08724.1 | - | 8 - 9 | 1084 | 73 | |
Y08725.1 | - | 8 - 9 | 1084 | 73 |