Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199.3 | NP_001190.1 | 4 - 5 | 817 | 65 |
NM_006129.4 | NP_006120.1 | 4 - 5 | 817 | 65 | |
XM_006716386.2 | XP_006716449.2 | 4 - 5 | 817 | 65 | |
XM_011544617.1 | XP_011542919.1 | 4 - 5 | 817 | 65 | |
XM_017013738.1 | XP_016869227.1 | 4 - 5 | 817 | 65 | |
GenBank mRNA | AB209590.1 | - | 4 - 5 | 770 | 65 |
AK291620.1 | - | 4 - 5 | 670 | 65 | |
BC101763.1 | - | 4 - 5 | 622 | 65 | |
BC101765.1 | - | 4 - 5 | 622 | 65 | |
BC136679.1 | - | 4 - 5 | 633 | 65 | |
BC142953.1 | - | 4 - 5 | 681 | 65 | |
M22488.1 | - | 4 - 5 | 580 | 65 | |
U50330.1 | - | 4 - 5 | 551 | 65 | |
Y08723.1 | - | 4 - 5 | 557 | 65 | |
Y08724.1 | - | 4 - 5 | 557 | 65 | |
Y08725.1 | - | 4 - 5 | 557 | 65 |